Gene Transcripts

Gene Symbol
MAGEF1
Entrez Gene ID
64110
Gene Name
MAGE family member F1
Gene Aliases
Chromosome Location
Chr. 3: 184710367 - 184712048 on Build GRCh38
UniGene ID
Hs.306123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022149.4 NP_071432.2 1 864
GenBank mRNA AF295378.1 - 1 816
AF320910.1 - 1 735
BC010056.2 - 1 821 3943384
BQ058163.1 - 1 470

View Full Product Details