Gene Transcripts

Gene Symbol
HERPUD2
Entrez Gene ID
64224
Gene Name
HERPUD family member 2
Gene Aliases
Chromosome Location
Chr. 7: 35632659 - 35695187 on Build GRCh38
UniGene ID
Hs.643051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022373.4 NP_071768.3 2 713
XM_006715765.1 XP_006715828.1 2 713
XM_011515473.1 XP_011513775.1 1 911
XM_017012506.1 XP_016867995.1 1 911
GenBank mRNA AK055708.1 - 1 860
AK315685.1 - 1 337
BC005091.1 - 2 519 3840918
BC020264.1 - 2 670 4878523

View Full Product Details