Gene Transcripts
- Gene Symbol
- NOM1
- Entrez Gene ID
- 64434
- Gene Name
- nucleolar protein with MIF4G domain 1
- Gene Aliases
- C7orf3, PPP1R113, SGD1
- Chromosome Location
- Chr. 7: 156949723 - 156973182 on Build GRCh38
- UniGene ID
- Hs.15825
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138400.1 |
NP_612409.1
|
4, 5 |
1654 |
|
GenBank mRNA |
AY576779.1 |
- |
4, 5 |
1654 |
|
View Full Product Details