Gene Transcripts

Gene Symbol
SPATS2
Entrez Gene ID
65244
Gene Name
spermatogenesis associated serine rich 2
Gene Aliases
Nbla00526, P59SCR, SCR59, SPATA10
Chromosome Location
Chr. 12: 49366905 - 49527441 on Build GRCh38
UniGene ID
Hs.654826
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293285.1 NP_001280214.1 15 2494
NM_001293286.1 NP_001280215.1 13 2347
NM_023071.3 NP_075559.2 14 2410
NR_120663.1 15 2454
XM_011538690.2 XP_011536992.1 15 1882
XM_011538691.2 XP_011536993.1 13 1777
GenBank mRNA AB075515.1 - 12 1438
AK023179.1 - 10 1319
AK023202.1 - 14 1809
AK024678.1 - 6 706
AK095582.1 - 15 2454
AK292334.1 - 13 1747
AL833614.1 - 15 1935
BC048299.1 - 13 1777 5520818
BX648916.1 - 13 1720

View Full Product Details