Gene Transcripts

Gene Symbol
TMEM53
Entrez Gene ID
79639
Gene Name
transmembrane protein 53
Gene Aliases
NET4
Chromosome Location
Chr. 1: 44653247 - 44674608 on Build GRCh38
UniGene ID
Hs.22157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300746.1 NP_001287675.1 2 1579
NM_001300747.1 NP_001287676.1 3 1611
NM_001300748.1 NP_001287677.1 3 1608
NM_024587.3 NP_078863.2 3 1701
XM_011542138.2 XP_011540440.1 3 1830
GenBank mRNA AL832539.1 - 2 1413
BC064520.1 - 3 1520 5582377

View Full Product Details