Gene Transcripts

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
Chromosome Location
Chr. 1: 48727523 - 48778603 on Build GRCh38
UniGene ID
Hs.475348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302082.1 NP_001289011.1 5 1186
NM_024603.3 NP_078879.2 5 1104
XM_011542141.2 XP_011540443.1 5 3604
XM_011542142.2 XP_011540444.1 4 1052
XM_017002331.1 XP_016857820.1 6 1137
XM_017002332.1 XP_016857821.1 6 1014
XM_017002334.1 XP_016857823.1 6 1233
GenBank mRNA AK021650.1 - 5 1048
BC007932.2 - 5 1056 4121760
BC011804.2 - 5 870 4336159
KU178776.1 - 5 966
KU178777.1 - 5 1048
KU178778.1 - 4 832

View Full Product Details