Gene Transcripts

Gene Symbol
C3orf52
Entrez Gene ID
79669
Gene Name
chromosome 3 open reading frame 52
Gene Aliases
TTMP
Chromosome Location
Chr. 3: 112086335 - 112131095 on Build GRCh38
UniGene ID
Hs.434247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024616.2 NP_078892.2 4 504
GenBank mRNA AK026839.1 - 4 448
AY830714.1 - 4 450
BC017064.1 - 4 437 3885219

View Full Product Details