Gene Transcripts

Gene Symbol
MYH14
Entrez Gene ID
79784
Gene Name
myosin, heavy chain 14, non-muscle
Gene Aliases
DFNA4, DFNA4A, FP17425, MHC16, MYH17, NMHC II-C, NMHC-II-C, PNMHH, myosin
Chromosome Location
Chr. 19: 50188186 - 50310544 on Build GRCh38
UniGene ID
Hs.467142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077186.1 NP_001070654.1 11 1215
NM_001145809.1 NP_001139281.1 11 1215
NM_024729.3 NP_079005.3 10 1191
XM_006723386.3 XP_006723449.1 11 1276
XM_011527320.2 XP_011525622.1 11 1293
XM_011527321.2 XP_011525623.1 10 1269
XM_011527323.2 XP_011525625.1 11 1283
GenBank mRNA AB290169.1 - 10 1185
AK125082.1 - 11 1294
AK307783.1 - 11 1198
AY165122.1 - 10 1191
BC018933.2 - 9 943
BC129803.1 - 11 1069

View Full Product Details