Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
79786
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
Chromosome Location
Chr. 16: 84648504 - 84667686 on Build GRCh38
UniGene ID
Hs.222731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024731.3 NP_079007.2 3 1255
XM_005256149.2 XP_005256206.1 3 1469
XM_017023674.1 XP_016879163.1 3 1217
XM_017023675.1 XP_016879164.1 3 1237
GenBank mRNA AK022605.1 - 2 1108
BC033821.1 - 3 1241 5219021

View Full Product Details