Gene Transcripts

Gene Symbol
PQLC1
Entrez Gene ID
80148
Gene Name
PQ loop repeat containing 1
Gene Aliases
Chromosome Location
Chr. 18: 79902420 - 79951665 on Build GRCh38
UniGene ID
Hs.288284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025078.4 NP_079354.2 4 527
XM_005266770.2 XP_005266827.1 4 539
XM_017025997.1 XP_016881486.1 5 614
XM_017025999.1 XP_016881488.1 5 614
XM_017026000.1 XP_016881489.1 5 614
XM_017026001.1 XP_016881490.1 3 544
XM_017026003.1 XP_016881492.1 5 614
XM_017026005.1 XP_016881494.1 2 178
GenBank mRNA AK123870.1 - 4 398
AK126188.1 - 2 1014
BC030140.2 - 4 491 4761671

View Full Product Details