Gene Transcripts

Gene Symbol
EGFL8
Entrez Gene ID
80864
Gene Name
EGF like domain multiple 8
Gene Aliases
C6orf8, NG3
Chromosome Location
Chr. 6: 32164605 - 32168285 on Build GRCh38
UniGene ID
Hs.731888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030652.3 NP_085155.1 3 292
NR_037860.1 3 316
GenBank mRNA AB190520.1 - 2 230
AK126457.1 - 1 1375
AK309657.1 - 1 974
AY358139.1 - 3 277
BC035574.1 - 3 269 5247785
BC052591.1 - 3 316 6295477

Gene Symbol
PPT2-EGFL8
Entrez Gene ID
100532746
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 6: 32153999 - 32168285 on Build GRCh38
UniGene ID
Hs.731888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037861.1 10 1728
GenBank mRNA AF020544.1 - 10 1882
AK307427.1 - 8 1181

View Full Product Details