Gene Transcripts

Gene Symbol
PTDSS2
Entrez Gene ID
81490
Gene Name
phosphatidylserine synthase 2
Gene Aliases
PSS2
Chromosome Location
Chr. 11: 448264 - 491399 on Build GRCh38
UniGene ID
Hs.731700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030783.1 NP_110410.1 10 1271
XM_005253153.1 XP_005253210.1 10 1252
XM_011520391.2 XP_011518693.1 10 1889
XM_011520392.2 XP_011518694.1 11 1268
XM_011520393.2 XP_011518695.1 9 986
XM_017018368.1 XP_016873857.1 12 3663
XM_017018369.1 XP_016873858.1 11 1393
XM_017018370.1 XP_016873859.1 9 1068
XM_017018371.1 XP_016873860.1 10 1182
GenBank mRNA AL834357.1 - 10 1185
BC001210.1 - 10 1271 3503123

View Full Product Details