Gene Transcripts

Gene Symbol
CFHR5
Entrez Gene ID
81494
Gene Name
complement factor H related 5
Gene Aliases
CFHL5, CFHR5D, FHR-5, FHR5
Chromosome Location
Chr. 1: 196975022 - 197009725 on Build GRCh38
UniGene ID
Hs.282594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030787.3 NP_110414.1 7 1239
XM_011510020.2 XP_011508322.1 7 1146
GenBank mRNA AF295327.1 - 7 1204
AK314026.1 - 7 1240
BC026282.1 - 7 1220
BC111773.1 - 7 1204 40034462

View Full Product Details