Gene Transcripts

Gene Symbol
OR2B2
Entrez Gene ID
81697
Gene Name
olfactory receptor family 2 subfamily B member 2
Gene Aliases
OR2B2Q, OR2B9, OR6-1, dJ193B12.4, hs6M1-10
Chromosome Location
Chr. 6: 27911185 - 27912396 on Build GRCh38
UniGene ID
Hs.307111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033057.2 NP_149046.2 1 293
GenBank mRNA BC136881.1 - 1 293
BC136901.1 - 1 293 9020892

View Full Product Details