Gene Transcripts

Gene Symbol
KRTAP1-3
Entrez Gene ID
81850
Gene Name
keratin associated protein 1-3
Gene Aliases
KAP1.2, KAP1.3, KAP1.6, KAP1.8A, KAP1.8B, KAP1.9, KRTAP1.3
Chromosome Location
Chr. 17: 41033884 - 41034855 on Build GRCh38
UniGene ID
Hs.534495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030966.1 NP_112228.1 1 784
GenBank mRNA AJ406927.1 - 1 784
BC069161.1 - 1 783 7216931
BC101150.1 - 1 778 40015405
BC101151.2 - 1 778 40015406
BC101152.1 - 1 808 40015409
BC101153.2 - 1 778 40015410

View Full Product Details