Gene Transcripts

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
Chromosome Location
Chr. 7: 99705037 - 99735200 on Build GRCh38
UniGene ID
Hs.111944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000765.4 NP_000756.3 8 840
GenBank mRNA BC067436.1 - 8 818
BX647143.1 - 5 1072
D00408.1 - 8 736

Gene Symbol
CYP3A7-CYP3A51P
Entrez Gene ID
100861540
Gene Name
CYP3A7-CYP3A51P readthrough
Gene Aliases
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L
Chromosome Location
Chr. 7: 99684679 - 99735200 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256497.2 NP_001243426.2 8 840
GenBank mRNA AF315325.1 - 8 733

View Full Product Details