Gene Transcripts

Gene Symbol
HDHD2
Entrez Gene ID
84064
Gene Name
haloacid dehalogenase like hydrolase domain containing 2
Gene Aliases
3110052N05Rik, HEL-S-301
Chromosome Location
Chr. 18: 47107402 - 47150520 on Build GRCh38
UniGene ID
Hs.465041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318765.1 NP_001305694.1 4, 5 579
NM_032124.4 NP_115500.1 4, 5 522
XM_005258371.4 XP_005258428.1 4, 5 542
XM_006722557.2 XP_006722620.1 5, 6 614
XM_011526227.2 XP_011524529.1 5, 6 598
XM_011526228.2 XP_011524530.1 5, 6 596
XM_017026039.1 XP_016881528.1 5, 6 635
XM_017026040.1 XP_016881529.1 5, 6 614
GenBank mRNA AK054590.1 - 4, 5 559
AK292098.1 - 4, 5 522
AL136681.1 - 4, 5 513
BC011894.2 - 4, 5 543 3944181
BC033031.1 - 4, 5 558 4793566
BC038100.2 - 4, 5 522 5266439
CR533487.1 - 3, 4 389
FJ224322.1 - 4, 5 513

View Full Product Details