Gene Transcripts

Gene Symbol
RAB6C
Entrez Gene ID
84084
Gene Name
RAB6C, member RAS oncogene family
Gene Aliases
WTH3
Chromosome Location
Chr. 2: 129979662 - 129982738 on Build GRCh38
UniGene ID
Hs.591552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032144.2 NP_115520.2 1 1302
GenBank mRNA AK055504.1 - 2 689
AL136727.1 - 1 1303
BC120999.1 - 1 1140 40116526

View Full Product Details