Gene Transcripts

Gene Symbol
HOPX
Entrez Gene ID
84525
Gene Name
HOP homeobox
Gene Aliases
CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
Chromosome Location
Chr. 4: 56647988 - 56681866 on Build GRCh38
UniGene ID
Hs.619396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145459.1 NP_001138931.1 3 1073
NM_001145460.1 NP_001138932.1 5 1557
NM_032495.5 NP_115884.4 4 1427
NM_139211.4 NP_631957.1 3 1029
NM_139212.3 NP_631958.1 3 1302
XM_017008728.1 XP_016864217.1 3 1064
XM_017008729.1 XP_016864218.1 3 946
XM_017008730.1 XP_016864219.1 3 994
XM_017008731.1 XP_016864220.1 3 1029
XM_017008732.1 XP_016864221.1 4 1305
XM_017008733.1 XP_016864222.1 3 869
XM_017008734.1 XP_016864223.1 3 925
GenBank mRNA AB059408.1 - 3 987
AB059409.1 - 3 870
AB059410.2 - 3 879
AF454763.1 - 3 893
AF492675.1 - 3 1278
AF492676.1 - 4 1010
AF492677.1 - 4 1054
AF492678.1 - 5 1184
AF492679.1 - 5 1140
AF492680.1 - 3 1030
AF492681.1 - 3 993
AK123318.1 - 1 3099
BC014225.2 - 2 1142 4335211

View Full Product Details