Gene Transcripts

Gene Symbol
TTBK1
Entrez Gene ID
84630
Gene Name
tau tubulin kinase 1
Gene Aliases
BDTK
Chromosome Location
Chr. 6: 43243484 - 43288259 on Build GRCh38
UniGene ID
Hs.485436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032538.1 NP_115927.1 14 3059
XM_011514947.2 XP_011513249.1 13 2873
XM_011514948.2 XP_011513250.1 2 3197
XM_017011364.1 XP_016866853.1 12 3110
XM_017011365.1 XP_016866854.1 12 3110
XM_017011366.1 XP_016866855.1 13 2849
XM_017011368.1 XP_016866857.1 2 3167
GenBank mRNA AB058758.1 - 12 2628
AB218664.1 - 15 2863

View Full Product Details