Gene Transcripts

Gene Symbol
TTBK1
Entrez Gene ID
84630
Gene Name
tau tubulin kinase 1
Gene Aliases
BDTK
Chromosome Location
Chr. 6: 43243484 - 43288259 on Build GRCh38
UniGene ID
Hs.485436
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032538.1 NP_115927.1 9, 10 1140
XM_011514947.2 XP_011513249.1 8, 9 930
XM_017011364.1 XP_016866853.1 8, 9 930
XM_017011365.1 XP_016866854.1 8, 9 930
XM_017011366.1 XP_016866855.1 8, 9 930
XM_017011367.1 XP_016866856.1 8, 9 930
GenBank mRNA AB058758.1 - 7, 8 709
AB218664.1 - 10, 11 944
AK131217.1 - 8, 9 870

View Full Product Details