Gene Transcripts

Gene Symbol
CCDC74A
Entrez Gene ID
90557
Gene Name
coiled-coil domain containing 74A
Gene Aliases
Chromosome Location
Chr. 2: 131522621 - 131533666 on Build GRCh38
UniGene ID
Hs.351461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258304.1 NP_001245233.1 4 844
NM_001258305.1 NP_001245234.1 4 718
NM_001258306.1 NP_001245235.1 4 520
NM_138770.2 NP_620125.1 4 718
XM_005263835.3 XP_005263892.1 4 618
XM_005263836.4 XP_005263893.1 2 1001
XM_005263837.1 XP_005263894.1 4 636
XM_006712828.3 XP_006712891.1 4 817
XM_017005234.1 XP_016860723.1 4 830
XM_017005235.1 XP_016860724.1 4 889
XM_017005236.1 XP_016860725.1 4 631
XM_017005237.1 XP_016860726.1 4 845
XM_017005238.1 XP_016860727.1 4 854
XM_017005239.1 XP_016860728.1 4 691
XM_017005240.1 XP_016860729.1 4 877
XM_017005241.1 XP_016860730.1 5 626
XM_017005242.1 XP_016860731.1 4 627
XM_017005243.1 XP_016860732.1 4 638
XM_017005244.1 XP_016860733.1 5 422
XM_017005245.1 XP_016860734.1 4 648
XM_017005246.1 XP_016860735.1 4 520
GenBank mRNA AB299086.1 - 4 759
AB299087.1 - 4 633
AK097664.1 - 2 1201
BC016861.1 - 4 633 3914851
BC063387.1 - 4 457 5494806
CV023784.1 - 4 581
DA375407.1 - 4 520

Gene Symbol
CCDC74B
Entrez Gene ID
91409
Gene Name
coiled-coil domain containing 74B
Gene Aliases
Chromosome Location
Chr. 2: 130139283 - 130145134 on Build GRCh38
UniGene ID
Hs.29383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258307.1 NP_001245236.1 4 521
NM_207310.2 NP_997193.1 4 719
XM_005263840.2 XP_005263897.1 4 744
XM_005263842.2 XP_005263899.1 4 546
XM_006712833.2 XP_006712896.1 4 756
XM_006712834.3 XP_006712897.1 4 696
XM_006712835.2 XP_006712898.1 4 558
XM_006712837.2 XP_006712900.1 4 498
XM_011512142.2 XP_011510444.1 2 987
GenBank mRNA AK057609.1 - 4 643
AK297246.1 - 3 1631
AL133619.1 - 1 1769
BC067771.1 - 4 466 4824929

View Full Product Details