Gene Transcripts

Gene Symbol
SEPT1
Entrez Gene ID
1731
Gene Name
septin 1
Gene Aliases
DIFF6, LARP, PNUTL3, SEP1
Chromosome Location
Chr. 16: 30378133 - 30382850 on Build GRCh38
UniGene ID
Hs.632176
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052838.4 NP_443070.5 7 667
XM_017022991.1 XP_016878480.1 7 647
GenBank mRNA AF085235.1 - 6 498
AK301045.1 - 5 593
AK303390.1 - 6 724
AL833004.1 - 2 340
AY034176.1 - 6 501
BC012161.1 - 5 489 4576220
BQ072902.1 - 7 680

View Full Product Details