Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040272.5 | NP_001035362.3 | 5 | 663 | |
NM_052866.4 | NP_443098.3 | 5 | 663 | ||
XM_011518063.2 | XP_011516365.1 | 7 | 801 | ||
XM_011518064.2 | XP_011516366.1 | 6 | 916 | ||
XM_017015310.1 | XP_016870799.1 | 6 | 7438 | ||
XM_017015311.1 | XP_016870800.1 | 7 | 801 | ||
XM_017015312.1 | XP_016870801.1 | 6 | 917 | ||
XM_017015313.1 | XP_016870802.1 | 6 | 564 | ||
XM_017015314.1 | XP_016870803.1 | 7 | 801 | ||
GenBank mRNA | AF176313.1 | - | 5 | 561 | |
AK290667.1 | - | 5 | 649 | ||
AK291956.1 | - | 5 | 603 | ||
AY358327.1 | - | 5 | 643 | ||
BC030262.1 | - | 5 | 575 | 5201199 | |
CX758410.1 | - | 5 | 562 |