Gene Transcripts

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
Chromosome Location
Chr. 11: 92224636 - 92896473 on Build GRCh38
UniGene ID
Hs.98523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008781.2 NP_001008781.2 8 4648
XM_017017178.1 XP_016872667.1 10 4833
XM_017017179.1 XP_016872668.1 11 5055
XM_017017180.1 XP_016872669.1 10 4832
XM_017017181.1 XP_016872670.1 9 4763
XM_017017182.1 XP_016872671.1 10 4832
XM_017017183.1 XP_016872672.1 9 4763
XM_017017184.1 XP_016872673.1 10 4831
XM_017017185.1 XP_016872674.1 9 2659
XM_017017186.1 XP_016872675.1 9 1947
XM_017017187.1 XP_016872676.1 9 6305

View Full Product Details