Gene Transcripts

Gene Symbol
WFIKKN2
Entrez Gene ID
124857
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2
Gene Aliases
GASP-1, WFDC20B, WFIKKNRP, hGASP-1
Chromosome Location
Chr. 17: 50834650 - 50842353 on Build GRCh38
UniGene ID
Hs.211475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175575.5 NP_783165.1 2 2163
XM_017024172.1 XP_016879661.1 2 1642
GenBank mRNA AF468657.1 - 2 1485
AK127743.1 - 2 1823
AK131196.1 - 733
AK298172.1 - 2 1642
AK312821.1 - 2 1657
AY358142.1 - 2 1998
BC140842.1 - 2 1557 9021710
BC140844.1 - 2 1557 9021712

View Full Product Details