Gene Transcripts

Gene Symbol
COPS9
Entrez Gene ID
150678
Gene Name
COP9 signalosome subunit 9
Gene Aliases
CSNAP, MYEOV2
Chromosome Location
Chr. 2: 240126548 - 240136802 on Build GRCh38
UniGene ID
Hs.293884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138336.1 NP_612209.1 4, 5 384
XM_017003411.1 XP_016858900.1 4, 5 809
GenBank mRNA AF453951.1 - 4, 5 384

View Full Product Details