Gene Transcripts

Gene Symbol
CPT1B
Entrez Gene ID
1375
Gene Name
carnitine palmitoyltransferase 1B
Gene Aliases
CPT1-M, CPT1M, CPTI, CPTI-M, M-CPT1, MCCPT1, MCPT1
Chromosome Location
Chr. 22: 50568861 - 50578667 on Build GRCh38
UniGene ID
Hs.439777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145134.1 NP_001138606.1 13 1708
NM_001145135.1 NP_001138607.1 14 1810
NM_001145137.1 NP_001138609.1 13 1834
NM_004377.3 NP_004368.1 14 1771
NM_152245.2 NP_689451.1 14 1810
NM_152246.2 NP_689452.1 14 1771
GenBank mRNA AK223174.1 - 14 1710
AK291316.1 - 14 1735
AK297866.1 - 13 1620
AK299395.1 - 14 1504
BC142954.1 - 13 1777 8860341
D87812.1 - 13 1691
U62733.1 - 13 1718
U66828.1 - 14 1710
Y08682.1 - 14 1723
Y08683.1 - 14 1723

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
Chromosome Location
Chr. 22: 50568861 - 50582999 on Build GRCh38
UniGene ID
Hs.439777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027928.2 24 3242
GenBank mRNA AB051457.1 - 21 3880
BC131570.1 - 16 2017 40108355

View Full Product Details