Gene Transcripts

Gene Symbol
KRTAP20-3
Entrez Gene ID
337985
Gene Name
keratin associated protein 20-3
Gene Aliases
KAP19D, KAP20.3, KRTAP19P4
Chromosome Location
Chr. 21: 30642864 - 30643136 on Build GRCh38
UniGene ID
Hs.580878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128077.1 NP_001121549.1 1 237
GenBank mRNA AB096958.1 - 1 237

View Full Product Details