Gene Transcripts

Gene Symbol
RBM33
Entrez Gene ID
155435
Gene Name
RNA binding motif protein 33
Gene Aliases
PRR8
Chromosome Location
Chr. 7: 155644494 - 155781485 on Build GRCh38
UniGene ID
Hs.591815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053043.2 NP_444271.2 17 3807
XM_005249531.1 XP_005249588.1 18 3975
XM_005249532.1 XP_005249589.1 18 3972
XM_005249533.1 XP_005249590.1 18 3945
XM_005249534.1 XP_005249591.1 17 3855
XM_005249535.1 XP_005249592.1 17 3825
XM_011515840.1 XP_011514142.1 17 3765
GenBank mRNA AK056415.1 - 2 215
AL832196.1 - 7 541
BC093947.1 - 3 313
BC093949.1 - 3 313
CN402759.1 - 4 669

View Full Product Details