Gene Transcripts

Gene Symbol
RASEF
Entrez Gene ID
158158
Gene Name
RAS and EF-hand domain containing
Gene Aliases
RAB45
Chromosome Location
Chr. 9: 82979585 - 83063177 on Build GRCh38
UniGene ID
Hs.129136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152573.3 NP_689786.2 8 1309
XM_005251730.3 XP_005251787.1 7 1289
XM_005251731.3 XP_005251788.1 8 1800
GenBank mRNA AK290986.1 - 8 1051
BC023566.1 - 8 1309 4138119

View Full Product Details