Gene Transcripts

Gene Symbol
LGI4
Entrez Gene ID
163175
Gene Name
leucine rich repeat LGI family member 4
Gene Aliases
LGIL3
Chromosome Location
Chr. 19: 35124513 - 35135274 on Build GRCh38
UniGene ID
Hs.65256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139284.2 NP_644813.1 4 963
XM_011526595.1 XP_011524897.1 5 444
XM_017026428.1 XP_016881917.1 4 292
XM_017026429.1 XP_016881918.1 5 364
XM_017026430.1 XP_016881919.1 3 146
GenBank mRNA AF467954.1 - 4 655
AJ487519.1 - 4 888
AJ487959.1 - 4 396
AL832968.1 - 2 104
AY358121.1 - 4 558
BC029943.1 - 5 428
BC087848.1 - 4 540
BC136694.1 - 4 438 9020684
BC136697.1 - 4 438 9020687

View Full Product Details