Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039876.2 | NP_001034965.1 | 6, 7 | 1082 | |
NM_001297735.2 | NP_001284664.1 | 4, 5 | 743 | ||
GenBank mRNA | AK093764.1 | - | 5, 6 | 2041 | |
BC052573.1 | - | 4, 5 | 733 | 6568945 |