Gene Transcripts

Gene Symbol
FAM47B
Entrez Gene ID
170062
Gene Name
family with sequence similarity 47 member B
Gene Aliases
Chromosome Location
Chr. X: 34942796 - 34944917 on Build GRCh38
UniGene ID
Hs.197801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152631.2 NP_689844.2 1 1544
GenBank mRNA AK093101.1 - 1 1526
BC035026.2 - 1 1544 4828379

View Full Product Details