Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024852.3 | NP_079128.2 | 16 | 2480 | |
NM_177422.2 | NP_803171.1 | 14 | 2222 | ||
XM_005270575.3 | XP_005270632.1 | 17 | 7934 | ||
XM_005270576.3 | XP_005270633.1 | 11 | 2718 | ||
XM_011540879.2 | XP_011539181.1 | 16 | 3362 | ||
XM_011540882.2 | XP_011539184.1 | 13 | 1577 | ||
XM_017000523.1 | XP_016856012.1 | 17 | 7950 | ||
XM_017000524.1 | XP_016856013.1 | 12 | 1748 | ||
XM_017000525.1 | XP_016856014.1 | 13 | 1582 | ||
XM_017000526.1 | XP_016856015.1 | 12 | 1541 | ||
XM_017000527.1 | XP_016856016.1 | 12 | 1720 | ||
XM_017000528.1 | XP_016856017.1 | 13 | 1669 | ||
GenBank mRNA | AK022827.1 | - | 16 | 2235 | |
AK292870.1 | - | 16 | 2446 | ||
AK302289.1 | - | 9 | 1229 | ||
AK303929.1 | - | 13 | 1764 | ||
BC025769.1 | - | 14 | 2046 |