Gene Transcripts
- Gene Symbol
- SDR16C5
- Entrez Gene ID
- 195814
- Gene Name
- short chain dehydrogenase/reductase family 16C, member 5
- Gene Aliases
- EPHD-2, RDH#2, RDH-E2, RDHE2, retSDR2
- Chromosome Location
- Chr. 8: 56299648 - 56320776 on Build GRCh38
- UniGene ID
- Hs.170673
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318049.1 |
NP_001304978.1
|
4, 5 |
1302 |
|
NM_001318050.1 |
NP_001304979.1
|
3, 4 |
1170 |
|
NM_138969.3 |
NP_620419.2
|
4, 5 |
1302 |
|
XM_011517479.1 |
XP_011515781.1
|
3, 4 |
1170 |
|
GenBank mRNA |
AB083038.1 |
- |
4, 5 |
1208 |
|
AK057667.1 |
- |
4, 5 |
644 |
|
AK095159.1 |
- |
4, 5 |
1302 |
|
AK294634.1 |
- |
3, 4 |
573 |
|
AY444559.1 |
- |
3, 4 |
569 |
|
BC037219.1 |
- |
4, 5 |
649 |
5181472 |
BC064525.1 |
- |
4, 5 |
621 |
5752917 |
View Full Product Details