Gene Transcripts

Gene Symbol
ZBTB7C
Entrez Gene ID
201501
Gene Name
zinc finger and BTB domain containing 7C
Gene Aliases
APM-1, APM1, ZBTB36, ZNF857C
Chromosome Location
Chr. 18: 48026886 - 48412639 on Build GRCh38
UniGene ID
Hs.515388
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039360.2 NP_001034449.1 2 916
NM_001318841.1 NP_001305770.1 4 1097
XM_005258229.4 XP_005258286.1 3 841
XM_011525861.2 XP_011524163.1 6 1321
XM_011525863.2 XP_011524165.1 5 1438
XM_011525864.2 XP_011524166.1 5 1193
XM_011525865.2 XP_011524167.1 5 1434
XM_011525866.2 XP_011524168.1 4 1179
XM_011525867.2 XP_011524169.1 4 968
XM_011525869.2 XP_011524171.1 5 1443
XM_011525870.2 XP_011524172.1 4 1139
XM_011525871.2 XP_011524173.1 2 1654
XM_017025605.1 XP_016881094.1 6 3116
XM_017025606.1 XP_016881095.1 6 1570
XM_017025607.1 XP_016881096.1 6 1325
XM_017025608.1 XP_016881097.1 5 2993
XM_017025609.1 XP_016881098.1 4 1310
GenBank mRNA AK296709.1 - 2 916
AM988836.1 - 2 904
AM988837.1 - 2 1049
AM988838.1 - 3 1099
AM988839.1 - 4 1097
EF120357.1 - 2 968

View Full Product Details