Gene Transcripts

Gene Symbol
OR5D18
Entrez Gene ID
219438
Gene Name
olfactory receptor family 5 subfamily D member 18
Gene Aliases
OR11-143
Chromosome Location
Chr. 11: 55819630 - 55820571 on Build GRCh38
UniGene ID
Hs.553624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001952.1 NP_001001952.1 1 257
GenBank mRNA BC136925.1 - 1 277
BC136927.1 - 1 280 9020919

View Full Product Details