Gene Transcripts
- Gene Symbol
- FAM13C
- Entrez Gene ID
- 220965
- Gene Name
- family with sequence similarity 13 member C
- Gene Aliases
- FAM13C1
- Chromosome Location
- Chr. 10: 59245933 - 59363186 on Build GRCh38
- UniGene ID
- Hs.607594
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001971.2 |
NP_001001971.1
|
9, 10 |
1164 |
|
NM_001143773.1 |
NP_001137245.1
|
12, 13 |
1629 |
|
NM_198215.3 |
NP_937858.2
|
11, 12 |
1458 |
|
XM_006717702.3 |
XP_006717765.1
|
13, 14 |
1459 |
|
XM_006717703.3 |
XP_006717766.1
|
13, 14 |
1641 |
|
XM_011539490.2 |
XP_011537792.1
|
12, 13 |
1514 |
|
XM_011539491.2 |
XP_011537793.1
|
13, 14 |
1560 |
|
XM_017015885.1 |
XP_016871374.1
|
12, 13 |
1372 |
|
XM_017015891.1 |
XP_016871380.1
|
10, 11 |
1335 |
|
XM_017015893.1 |
XP_016871382.1
|
12, 13 |
1436 |
|
GenBank mRNA |
AK289796.1 |
- |
11, 12 |
1384 |
|
AK294805.1 |
- |
11, 12 |
1373 |
|
AK299924.1 |
- |
12, 13 |
1629 |
|
BC036453.1 |
- |
11, 12 |
1416 |
5270033 |
BC064431.1 |
- |
9, 10 |
1164 |
5764753 |
View Full Product Details