Gene Transcripts

Gene Symbol
LRRC57
Entrez Gene ID
255252
Gene Name
leucine rich repeat containing 57
Gene Aliases
Chromosome Location
Chr. 15: 42528332 - 42548804 on Build GRCh38
UniGene ID
Hs.234681
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153260.2 NP_694992.2 4, 5 619
XM_011521423.2 XP_011519725.1 3, 4 507
XM_011521424.2 XP_011519726.1 3, 4 507
GenBank mRNA AK094131.1 - 4, 5 613
AK094891.1 - 3, 4 853
AK309861.1 - 3, 4 1184
BC058935.1 - 4, 5 575 5578024
BX538324.1 - 3, 4 765
CN350663.1 - 1, 2 83
CR749223.1 - 4, 5 588

View Full Product Details