Gene Transcripts

Gene Symbol
NUTM1
Entrez Gene ID
256646
Gene Name
NUT midline carcinoma family member 1
Gene Aliases
C15orf55, FAM22H, NUT
Chromosome Location
Chr. 15: 34343315 - 34357737 on Build GRCh38
UniGene ID
Hs.525769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284292.1 NP_001271221.1 4 1301
NM_001284293.1 NP_001271222.1 3 1207
NM_175741.2 NP_786883.1 4 1386
GenBank mRNA AF482429.1 - 3 990
AK302656.1 - 4 1301
AK302680.1 - 3 944
BC114518.1 - 3 962 40036588
BC114567.1 - 3 957 40036589

View Full Product Details