Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320747.1 | NP_001307676.1 | 12 | 1568 | |
NM_001988.3 | NP_001979.2 | 12 | 1568 | ||
GenBank mRNA | AK290333.1 | - | 12 | 1568 | |
BC126103.1 | - | 12 | 1347 | 8991819 | |
BC126105.1 | - | 12 | 1347 | 8991821 | |
BC143814.1 | - | 12 | 1347 | 9052329 | |
U53786.1 | - | 12 | 1412 |