Gene Transcripts

Gene Symbol
F9
Entrez Gene ID
2158
Gene Name
coagulation factor IX
Gene Aliases
F9 p22, FIX, HEMB, P19, PTC, THPH8
Chromosome Location
Chr. X: 139530720 - 139563459 on Build GRCh38
UniGene ID
Hs.522798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000133.3 NP_000124.1 8 1001
NM_001313913.1 NP_001300842.1 7 887
XM_005262397.4 XP_005262454.1 7 888
GenBank mRNA A13997.1 - 8 973
AB186358.1 - 7 858
AK292749.1 - 8 972
BC109214.1 - 8 1001 40007636
BC109215.1 - 8 1001 40007637
FR846238.1 - 8 972
FR846239.1 - 8 972
FR846240.1 - 8 972
J00136.1 - 8 975
J00137.1 - 8 973
M11309.1 - 8 972

View Full Product Details