Gene Transcripts

Gene Symbol
SLC25A34
Entrez Gene ID
284723
Gene Name
solute carrier family 25 member 34
Gene Aliases
Chromosome Location
Chr. 1: 15735088 - 15741392 on Build GRCh38
UniGene ID
Hs.631867
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207348.2 NP_997231.1 4 872
XM_011541292.2 XP_011539594.1 4 2098
XM_011541293.1 XP_011539595.1 4 2098
GenBank mRNA BC027998.1 - 4 872
CR749264.1 - 1 109

View Full Product Details