Gene Transcripts
- Gene Symbol
- SLC36A3
- Entrez Gene ID
- 285641
- Gene Name
- solute carrier family 36 member 3
- Gene Aliases
- PAT3, TRAMD2, tramdorin2
- Chromosome Location
- Chr. 5: 151276365 - 151303773 on Build GRCh38
- UniGene ID
- Hs.720893
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145017.1 |
NP_001138489.1
|
1, 2 |
552 |
|
NM_181774.3 |
NP_861439.3
|
1, 2 |
552 |
|
XM_011537627.2 |
XP_011535929.1
|
1, 2 |
552 |
|
XM_011537629.2 |
XP_011535931.1
|
1, 2 |
552 |
|
XM_011537634.2 |
XP_011535936.1
|
1, 2 |
552 |
|
GenBank mRNA |
AK127978.1 |
- |
1, 2 |
545 |
|
AY162215.1 |
- |
1, 2 |
370 |
|
BC101092.1 |
- |
1, 2 |
275 |
40009824 |
BC101093.1 |
- |
1, 2 |
275 |
40009826 |
BC101094.1 |
- |
1, 2 |
275 |
40009832 |
BC101095.2 |
- |
1, 2 |
275 |
40009834 |
View Full Product Details