Gene Transcripts

Gene Symbol
NPEPPS
Entrez Gene ID
9520
Gene Name
aminopeptidase puromycin sensitive
Gene Aliases
AAP-S, MP100, PSA
Chromosome Location
Chr. 17: 47522950 - 47623276 on Build GRCh38
UniGene ID
Hs.443837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006310.3 NP_006301.3 21 2736
XM_011525496.1 XP_011523798.1 22 2538
XM_017025372.1 XP_016880861.1 21 2698
XM_017025373.1 XP_016880862.1 22 2538
GenBank mRNA AJ132583.1 - 21 2466
AK096491.1 - 6 1194
AK096709.1 - 23 2523
AK293450.1 - 15 1668
AK293995.1 - 14 1650
AK296887.1 - 20 2379
AK297107.1 - 20 2240
AK303037.1 - 22 2531
BC065294.1 - 21 2425 6059589

View Full Product Details