Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 13 | 2318 | |
XM_005257253.1 | XP_005257310.1 | 13 | 2363 | ||
GenBank mRNA | AF055008.1 | - | 13 | 2124 | |
AK000607.1 | - | 13 | 2102 | ||
AK023348.1 | - | 10 | 1606 | ||
AK222522.1 | - | 13 | 2102 | ||
AY124489.1 | - | 12 | 2086 | ||
BC000324.2 | - | 12 | 1816 | 2821810 | |
BC010577.2 | - | 12 | 2072 | 3457813 | |
X62320.1 | - | 13 | 2102 |