Gene Transcripts

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
Chromosome Location
Chr. 16: 81314941 - 81380198 on Build GRCh38
UniGene ID
Hs.112569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022041.3 NP_071324.1 3 443
XM_017023734.1 XP_016879223.1 2 353
GenBank mRNA AF291673.1 - 3 439
AK095378.1 - 3 443
BC044840.1 - 3 430 2900666
KU178409.1 - 3 295

View Full Product Details