Gene Transcripts

Gene Symbol
NLRP10
Entrez Gene ID
338322
Gene Name
NLR family pyrin domain containing 10
Gene Aliases
CLR11.1, NALP10, NOD8, PAN5, PYNOD
Chromosome Location
Chr. 11: 7959023 - 7965427 on Build GRCh38
UniGene ID
Hs.449636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176821.3 NP_789791.1 2 1035
XM_011520043.2 XP_011518345.1 3 1245
GenBank mRNA AY154465.1 - 2 1018
AY489192.1 - 2 1035
BC104957.1 - 2 1112 8143960

View Full Product Details