Gene Transcripts
- Gene Symbol
- OSTN
- Entrez Gene ID
- 344901
- Gene Name
- osteocrin
- Gene Aliases
- MUSCLIN
- Chromosome Location
- Chr. 3: 191199060 - 191265601 on Build GRCh38
- UniGene ID
- Hs.526794
- Species
- Homo sapiens
- Gene Symbol
- OSTN-AS1
- Entrez Gene ID
- 106480738
- Gene Name
- OSTN antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 191213291 - 191234605 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BQ001512.1 |
- |
3 |
205 |
|
View Full Product Details